missing translation for 'onlineSavingsMsg'
Learn More

Chlorsulfuron in Acetonitrile 100μg/mL, Fisher Chemical™

Product Code. 12993224
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 100μg/mL
CAS 64902-72-3
Packaging Ampoule
Quantity 1 mL
Molecular Formula C12H12ClN5O4S
MDL Number MFCD00128059
UN Number 1648
InChI Key VJYIFXVZLXQVHO-UHFFFAOYSA-N
SMILES COC1=NC(C)=NC(NC(=O)NS(=O)(=O)C2=CC=CC=C2Cl)=N1
IUPAC Name 1-(2-chlorophenyl)sulfonyl-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)urea
Molecular Weight (g/mol) 357.77
PubChem CID 47491
ChEBI CHEBI:47066
Solvent or Matrix Acetonitrile
Chemical Name or Material Chlorsulfuron
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Chlorsulfuron 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'