missing translation for 'onlineSavingsMsg'
Learn More

Diquat dibromide hydrate in Water 10μg/mL, Fisher Chemical™

Product Code. 12926593
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule(s)
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 10μg/mL
CAS 6385-62-2
Packaging Ampoule(s)
Quantity 10 mL
Molecular Formula C12H14Br2N2O
MDL Number MFCD00211319
InChI Key KLJOSQAQZMLLMB-UHFFFAOYSA-L
SMILES O.[Br-].[Br-].C1C[N+]2=CC=CC=C2C2=CC=CC=[N+]12
IUPAC Name 6,7-dihydrodipyrido[1,2-b
Molecular Weight (g/mol) 362.07
PubChem CID 115268
Solvent or Matrix Water
Chemical Name or Material Diquat dibromide hydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'