missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metabolite Endosulfan-total (sulfate) in Methanol 100μg/mL, Fisher Chemical™
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule(s)
Specifications
Concentration | 100μg/mL |
CAS | 1031-07-8 |
Packaging | Ampoule(s) |
Quantity | 1 mL |
Molecular Formula | C9H6Cl6O4S |
InChI Key | AAPVQEMYVNZIOO-WINLOITPSA-N |
SMILES | C1C2C(COS(=O)(=O)O1)C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl |
Molecular Weight (g/mol) | 422.903 |
PubChem CID | 71312398 |
Solvent or Matrix | Methanol |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'