missing translation for 'onlineSavingsMsg'
Learn More

Heptachlor-exo-epoxide in Methanol 10μg/mL, Fisher Chemical™

Product Code. 12926853
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule(s)
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 10μg/mL
CAS 1024-57-3
Packaging Ampoule(s)
Quantity 10 mL
Molecular Formula C10H5Cl7O
InChI Key ZXFXBSWRVIQKOD-GOWUZIGHSA-N
SMILES C12C(C(C3C1O3)Cl)C4(C(=C(C2(C4(Cl)Cl)Cl)Cl)Cl)Cl
Molecular Weight (g/mol) 389.299
PubChem CID 94865622
Solvent or Matrix Methanol
Chemical Name or Material Heptachlor-exo-epoxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'