missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, bis-butyl ester in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12956124
Click to view available options
Quantity:
1 mL
Packaging:
Ampoule
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 100μg/mL
CAS 84-74-2
Packaging Ampoule
Quantity 1 mL
Type Single-component Standard
Molecular Formula C16H22O4
MDL Number MFCD00009441
UN Number 1230
InChI Key DOIRQSBPFJWKBE-UHFFFAOYSA-N
SMILES CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC
IUPAC Name dibutyl benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 278.35
PubChem CID 3026
ChEBI CHEBI:34687
Solvent or Matrix Methanol
Chemical Name or Material Phthalic acid, bis-butyl ester
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution Phthalic acid,bis-butyl ester 100ug/ml in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'