missing translation for 'onlineSavingsMsg'
Learn More
Terbumeton-desethyl in Methanol 10μg/mL, Fisher Chemical™ Product Code.: 12970114

Terbumeton-desethyl in Methanol 10μg/mL, Fisher Chemical™

Product Code. 12970114
10 mL, Ampoule
Click to view available options
Quantity:
10 mL
Packaging:
Ampoule
This item is not returnable. View return policy

Product Code. 12970114

Brand: Fisher Chemical P852730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 10μg/mL
CAS 30125-64-5
Packaging Ampoule
Quantity 10 mL
Type Organic Single-component Standard Solution
Molecular Formula C8H15N5O
InChI Key HSYISQLUXXBNFW-UHFFFAOYSA-N
SMILES CC(C)(C)NC1=NC(=NC(=N1)N)OC
IUPAC Name 2-N-tert-butyl-6-methoxy-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol) 197.242
PubChem CID 185719
Solvent or Matrix Methanol
Physical Form Liquid
Chemical Name or Material Terbumeton-desethyl
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Terbumeton-desethyl in Methanol 10μg/mL, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.