Learn More
Sampling Systems™ Metal Detectable Scrapers
Metal Detectable Scrapers are ideal for working with powders, pastes, gels and granules in production, industrial and laboratories.
Brand: Sampling Systems™ M1019B-110
Additional Details : Weight : 0.90000kg
Description
- They are made from a special chemically resistant grade of PP.
- The whole range is manufactured in cleanroom conditions and packed in a class 7 (10,000) cleanroom.
- This ensures the cleanliness of the product.
- The robust design means they can be used for years and years or used as a single use product straight from the bag.
- Simply unwrap and use.
- Our ergonomic handle means they are easy to use.
- Made from high quality PP.
- Conforms to the latest FDA and EU food requirements.
- 110 mm blade width is a standard size.
Specifications
Blue | |
Polypropylene | |
110 mm |