Learn More
Sampling Systems™ Metal Detectable Spoons
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Pack of 4
Description
- Our Metal Detectable Spoons are made from food grade polypropylene (PP).
- A special food grade filler is mixed with the PP and this makes the final product both metal detectable and visible by X-rays.
- All products in this metal detectable range are moulded and packed in a cleanroom.
- This ensures the cleanliness of the product.
- Simply unwrap and use.
- They are ideal for use with powders, granules, liquids and pastes.
- The range can be easily cleaned after use.
- There are no dead spaces or crevices which can lead to bacteria traps.
- Made from high quality PP.
- Conforms to latest EU food and FDA requirements.
- Just a small part of metal detectable range of products.
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| Color | Blue |
| Material | Polypropylene |