Learn More
Astell Scientific™ Nelldorn Glass Dryer
An economical and efficient method of drying and storing most containers, glassware and plasticware.
Brand:  Astell Scientific™ DCU012
Product Code. 12753015
  • £2105.00 / Each
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unit can be used for most types of dispensing containers including plastics
  • Equipment can be left on the rack until drying is complete
  • Capable of holding up to 60 items with 15-20 minutes drying times
  • Wall mountable or free-standing
  • Thermostatically controlled
  • Economical, quick and efficient
  • Comes with a variable 120-minute timer
  • A large volume of air is drawn over the controlled heating elements and forced through each drying tube
  • Rapid flow of air dries glass or plastic quickly and without any damage to the apparatus
  • Very low power consumption
  • Stepped design allows the removal of any item without disturbing the remainder of the load and facilitates the run-off of water to the front of the unit where a small drain pipe prevents it collecting at the base
  • Air intake is recessed at the back of the unit to allow wall or bench mounting
  • Fitted with 60 rigid stain resistant nylon drying tubes with an adjustable timer which can be preset to switch off at your desired time
Specifications
Specifications
Drying Rack
Variable 120 minute timer
490 mm
50 Hz
Can Hold 30 x 75 mm Tubes, 13 x 100 mm Tubes, 9 x 40 mm Tubes, and 8 x 152 mm Tubes
430 mm
220/240 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Astell Scientific™ Nelldorn Glass Dryer > Dimensions: 190D x 490W x 430mmH