Learn More
Brady™ Metal Wall Lock Box
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Metal Wall Lock Box is made of durable, powder-coated steel with key slot on top. Includes clear Lexan window that slides out when locks are removed.
£105.00
Specifications
Product Type | Wall Mount Lock Box |
---|---|
Color | Yellow on Red |
Material | Powder-coated Steel |
Displayed Text | GROUP LOCK BOX |
Height (English) | 7 in. |
Description
- For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
- Reduces the number of locks being used
- Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
- Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Wall Mount Lock Box | |
Powder-coated Steel | |
7 in. |
Yellow on Red | |
GROUP LOCK BOX | |
1 Ea. |