Description
- Comfortable spatula temples with soft tips for a perfect fit
- Soft adjustable nose pads to custom fit virtually all face shapes and sizes
- Optical class 1 for maximum clarity
- Anti-Scratch and Anti-Fog coating for increased durability and improved vision
- Soft-touch temple tips provide additional comfort
Specifications
Specifications
Certifications/Compliance | CE Approved, EN 166: 2001 |
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety glasses |
Lens Material | Polycarbonate |
Frame Material | Metal |
Product Content Correction
Product Title
3M™ Metaliks™ Safety Glasses >
Spot an opportunity for improvement?Share a Content Correction