Learn More
3M™ Metaliks™ Sport Safety Glasses
Product Code. 15799667
Click to view available options
Lens Tint:
Amber
Blue, Mirror
3M™ Metaliks™ Sport Safety Glasses have a semi-rimless metal frame and optimal field of vision.
  • Semi-rimless metal frame and optimal field of vision
  • Soft-touch temple tips provide additional comfort
  • Soft saddle nose pad for enhanced comfort
  • Optical class 1 for maximum clarity
  • Anti-Scratch and Anti-Fog coating for increased durability and improved vision
  • Soft-touch temple tips provide additional comfort
  • Blue Mirror coating for increased durability and improved vision
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166: 2001
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Amber
Product Type Safety glasses
For Use With (Application) Construction, forestry
Frame Color Silver
Lens Material Polycarbonate
Frame Material Metal