missing translation for 'onlineSavingsMsg'
Learn More
Metaphosphoric acid, 39-43%, bal. NaPO{3} (Stabilizer), Thermo Scientific Chemicals Product Code.: 11419986

Metaphosphoric acid, 39-43%, bal. NaPO{3} (Stabilizer), Thermo Scientific Chemicals

Product Code. 11419986
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11419986

Brand: Thermo Scientific Alfa Aesar B20932.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Metaphosphoric acid is used as a dehydrating agent. Its derivative, sodium hexametaphosphate is used as a sequestrant and a food additive. It is also used as a phosphorylating agent, analytical reagent and in dental cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Metaphosphoric acid is used as a dehydrating agent. Its derivative, sodium hexametaphosphate is used as a sequestrant and a food additive. It is also used as a phosphorylating agent, analytical reagent and in dental cements.

Solubility
Soluble in alcohol.

Notes
Form: Lumps.Hygroscopic. Moisture sensitive. Incompatible with strong bases, nitromethane and metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37267-86-0
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Density 2.2
Sensitivity Hygroscopic
Quantity 500 g
UN Number UN3260
Merck Index 14,7345
Solubility Information Soluble in alcohol.
Formula Weight 79.98
Percent Purity 39 to 43%
Assay bal. NaPO3 (Stabilizer)
Chemical Name or Material Metaphosphoric Acid
compliance-icons
Product Identifier
  • Metaphosphoric acid, 39-43%, balance NaPO3 (Stabilizer)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Metaphosphoric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Metaphosphoric acid, 39-43%, bal. NaPO{3} (Stabilizer), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.