missing translation for 'onlineSavingsMsg'
Learn More
Metaphosphoric acid, ACS, 33.5-36.5%, bal. NaPO{3} (Stablizer), Thermo Scientific Chemicals Product Code.: 11377578

Metaphosphoric acid, ACS, 33.5-36.5%, bal. NaPO{3} (Stablizer), Thermo Scientific Chemicals

Product Code. 11377578
500 g
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11377578

Brand: Thermo Scientific Alfa Aesar 033267.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Metaphosphoric acid is used as a dehydrating agent. Its derivative, sodium hexametaphosphate is used as a sequestrant and a food additive. It is also used as phosphorylating agent, analytical reagent and in dental cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37267-86-0
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonateShow MoreShow Less
PubChem CID 3084658
SMILES OP(=O)=O

Specifications

Appearance White crystalline
Color White
Density 2.2 g/cm3 at 20&de g;C
Sensitivity Hygroscopic
Quantity 500 g
Physical Form Crystalline
UN Number 3260
Solubility Information Decomposes in water. Soluble in alcohol.
Formula Weight 79.98
Grade ACS
Chemical Name or Material Metaphosphoric Acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252725 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number033267.09,033267.22,033267.36
Lot Number10252723 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number033267.09,033267.22,033267.36
Lot Number10252721 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number033267.09,033267.22,033267.36
Lot Number10252719 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number033267.09,033267.22,033267.36
Lot Number10252724 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number033267.09,033267.22,033267.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Metaphosphoric acid, ACS, 33.5-36.5%, bal. NaPO{3} (Stablizer), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.