missing translation for 'onlineSavingsMsg'
Learn More
MetFormin hydrochloride, 98%, For HPLC analysis, MP Biomedicals™ Product Code.: 11482432

MetFormin hydrochloride, 98%, For HPLC analysis, MP Biomedicals™

Product Code. 11482432 Shop All MP Biomedicals Products
10 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 11482432

Brand: MP Biomedicals 0215169110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1115-70-4
Molecular Formula C4H12ClN5
Molecular Weight (g/mol) 165.63
MDL Number MFCD00012582
InChI Key OETHQSJEHLVLGH-UHFFFAOYSA-N
Synonym metformin hydrochloride, 1,1-dimethylbiguanide hydrochloride, metformin hcl, glucophage, riomet, diabefagos, meguan, metformin monohydrochloride, benofomin, denkaform
PubChem CID 14219
IUPAC Name hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride
SMILES [H+].[Cl-].CN(C)C(=N)N=C(N)N

Specifications

Color White
Quantity 10 g
Formula Weight 165.6
Percent Purity 0.98
Physical Form Crystalline Powder
Chemical Name or Material Metformin hydrochloride, For HPLC analysis
compliance-icons
Product Identifier
  • Metformin Hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Metformin Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MetFormin hydrochloride, 98%, For HPLC analysis, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.