missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methacholine chloride, 100.4%, MP Biomedicals™
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Specifications
Specifications
Color | White |
Assay Percent Range | ≥98% |
Quantity | 25 g |
Formula Weight | 195.7 |
Physical Form | Powder |
Chemical Name or Material | Methacholine chloride |
Safety and Handling
Product Identifier
- Methacholine Chloride
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : methacholine chloride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'