Learn More
Methacrylic acid, 99.5%, extra pure, stabilized, Thermo Scientific Chemicals
£17.00 - £533.00
Chemical Identifiers
CAS | 79-41-4, 150-76-5 |
---|---|
Molecular Weight (g/mol) | 86.09 |
MDL Number | MFCD00002651 |
InChI Key | CERQOIWHTDAKMF-UHFFFAOYSA-N |
Synonym | methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl |
PubChem CID | 4093 |
ChEBI | CHEBI:25219 |
IUPAC Name | 2-methylprop-2-enoic acid |
SMILES | CC(=C)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10685412
|
Thermo Scientific Acros
168310250 |
25 mL | Glass bottle |
£17.00
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10782781
|
Thermo Scientific Acros
168315000 |
500 mL | Glass bottle |
£43.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10184573
|
Thermo Scientific Acros
168310251 |
25 L | Metal Drum |
£533.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79-41-4, 150-76-5 | |
MFCD00002651 | |
methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl | |
CHEBI:25219 | |
CC(=C)C(=O)O |
86.09 | |
CERQOIWHTDAKMF-UHFFFAOYSA-N | |
4093 | |
2-methylprop-2-enoic acid |
Specifications
79-41-4, 150-76-5 | |
1.0100g/mL | |
65°C | |
99.4% min. (GC) | |
H2C=C(CH3)CO2H | |
MFCD00002651 | |
15, 6013 | |
methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl | |
CERQOIWHTDAKMF-UHFFFAOYSA-N | |
2-methylprop-2-enoic acid | |
86.09 | |
CHEBI:25219 | |
86.09 | |
Extra Pure | |
Methacrylic acid, Stabilized |
16.0°C | |
163.0°C | |
Authentic | |
C4H6O2 | |
1.4295 to 1.4315 | |
02, 421 | |
1.01 | |
Solubility in water: 97g/L (20°C). Other solubilities: miscible with alcohol,ether,esters,ketones | |
CC(=C)C(=O)O | |
250ppm (ca.) MEHQ | |
4093 | |
1.4 mPa.s (20°C) | |
99.5% | |
Liquid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin th
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-204-4
- Methacrylic acid, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only