missing translation for 'onlineSavingsMsg'
Learn More

Methacycline hydrochloride, MedChemExpress

Product Code. p-200100998
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16438316

Brand: Medchem Express HYB0449

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Methacycline hydrochloride is a tetracycline antibiotic and can inhibits bacterial protein synthesis. Methacycline hydrochloride is a potent epithelial-mesenchymal transition (EMT) inhibitor. Methacycline hydrochloride blocks EMT in vitro and fibrogenesis in vivo without directly affecting TGF-β1 Smad signaling. Methacycline hydrochloride is an antimicrobial and has the potential for pulmonary fibrosis.

Methacycline hydrochloride is a tetracycline antibiotic and can inhibits bacterial protein synthesis. Methacycline hydrochloride is a potent epithelial-mesenchymal transition (EMT) inhibitor. Methacycline hydrochloride blocks EMT in vitro and fibrogenesis in vivo without directly affecting TGF-β1 Smad signaling. Methacycline hydrochloride is an antimicrobial and has the potential for pulmonary fibrosis.

Specifications

Chemical Name or Material Methacycline hydrochloride
CAS 3963-95-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H23ClN2O8
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (104.41 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (13.93 mM; Need ultrasonic)
SMILES O[C@@](C(O)=C(C(C1=C2C=CC=C1O)=O)[C@](C2=C)([H])[C@@H]3O)(C(C(C(N)=O)=C4O)=O)[C@]3([H])[C@@H]4N(C)C.Cl
Molecular Weight (g/mol) 478.88
Formula Weight 478.88
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.