Learn More
Methanesulfonic acid, pure, 70% solution in water, Thermo Scientific Chemicals
£22.41 - £202.01
Chemical Identifiers
CAS | 75-75-2 |
---|---|
Molecular Formula | CH4O3S |
Molecular Weight (g/mol) | 96.1 |
MDL Number | MFCD00007518 |
InChI Key | AFVFQIVMOAPDHO-UHFFFAOYSA-N |
Synonym | methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane |
PubChem CID | 6395 |
ChEBI | CHEBI:27376 |
IUPAC Name | methanesulfonic acid |
SMILES | CS(=O)(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10646132
|
Thermo Scientific Acros
125622500 |
250 mL | Glass Bottle |
£22.41
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10235030
|
Thermo Scientific Acros
125620010 |
1 L | Glass Bottle |
£56.98
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10455555
|
Thermo Scientific Acros
125620050 |
5 L | Plastic bottle |
£202.01
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-75-2 | |
96.1 | |
AFVFQIVMOAPDHO-UHFFFAOYSA-N | |
6395 | |
methanesulfonic acid |
CH4O3S | |
MFCD00007518 | |
methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane | |
CHEBI:27376 | |
CS(=O)(=O)O |
Specifications
75-75-2 | |
1.3500g/mL | |
CH4O3S | |
1.4130 to 1.4150 | |
04, 4 | |
15, 6026 | |
methylsulfonic acid, methanesulphonic acid, methanesulfonicacid, kyselina methansulfonova, methansulfonsaeure, ccris 2783, kyselina methansulfonova czech, ch3so3h, methane sulfonic acid, sulfomethane | |
AFVFQIVMOAPDHO-UHFFFAOYSA-N | |
methanesulfonic acid | |
6395 | |
96.1 | |
Pure | |
Methanesulfonic acid, 70% Solution in Water |
-54.0°C | |
>110°C | |
CH3SO3H | |
MFCD00007518 | |
01,666; 02,270; 04,326; 10,256; 11,321; 12,212; 13,176 | |
1.35 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether,insoluble in hexane and methylcyclopentane,1.50wt% benzene- 0.23wt% in o-chlorotoluene (26°C),0.38wt% toluene- 0.47wt% in ethyl disulfide (26&de | |
CS(=O)(=O)O | |
96.1 | |
CHEBI:27376 | |
69.5 to 70.5% | |
Solution |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
D
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-898-6
- Methanesulfonic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.