missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic Acid, Sodium Salt, Ion Pair Chromatography, Fisher Chemical

Product Code. 10458410 Shop All Fisher Chemical Products
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 10458410

Brand: Fisher Chemical M/3876/46

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 2386-57-4
Color Cream to White
Physical Form Solid
Quantity 25 g
Molecular Formula CH3NaO3S
MDL Number 64389
Synonym sodium methanesulfonate, methanesulfonic acid sodium salt, methanesulfonic acid, sodium salt, sodium methanesulphonate, sodium mesylate, methylsulfonic acid sodium salt, 75-75-2 parent, sodium methyl sulfonate, methanesulfonic acid, sodium salt 1:1, sodium methylsulfonate
InChI Key KKVTYAVXTDIPAP-UHFFFAOYSA-M
SMILES CS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;methanesulfonate
Molecular Weight (g/mol) 118.082
PubChem CID 638112
Formula Weight 118.09g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Methanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.