missing translation for 'onlineSavingsMsg'
Learn More
Methanesulfonyl chloride, 99.5%, Thermo Scientific Chemicals Product Code.: 10735961

Methanesulfonyl chloride, 99.5%, Thermo Scientific Chemicals

Product Code. 10735961
100 mL, Glass bottle
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10735961

Brand: Thermo Scientific Acros 125641000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-63-0
Molecular Formula CH3ClO2S
Molecular Weight (g/mol) 114.54
MDL Number MFCD00007454
InChI Key QARBMVPHQWIHKH-UHFFFAOYSA-N
Synonym mesyl chloride, methylsulfonyl chloride, methyl sulfochloride, methanesulphonyl chloride, chloro methyl sulfone, methanesulfuryl chloride, methanesulfonic acid chloride, methyl sulfonyl chloride, chloromethyl sulfone, mesylchloride
PubChem CID 31297
IUPAC Name methanesulfonyl chloride
SMILES CS(Cl)(=O)=O

Specifications

Melting Point -33.0°C
Density 1.4800g/mL
Boiling Point 161.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Linear Formula CH3SO2Cl
Refractive Index 1.4435 to 1.4455
Quantity 100 mL
Beilstein 04,5
Fieser 01,662; 02,268; 04,326; 05,435; 06,362; 07,225; 11,322; 13,176; 17,180
Merck Index 15,6027
Specific Gravity 1.48
Solubility Information Solubility in water: insoluble (reacts with water). Other solubilities: soluble in ethanol, ether, acetone and other, common organic solvents
Viscosity 1.97 mPa.s (25°C)
Formula Weight 114.55
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material Methanesulfonyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0449340 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number125640025,125645000,125640010,125641000,125642500
Lot NumberA0457353 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number125640025,125645000,125640010,125641000,125642500
Lot NumberA0469446 Certificate TypeCertificate of Analysis Date24/01/2025 Catalog Number125640025,125645000,125640010,125641000,125642500
Lot NumberA0466190 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog Number125640025,125645000,125640010,125641000,125642500
Lot NumberA0457353 Certificate TypeCertificate of Analysis Date06/05/2024 Catalog Number125640025,125645000,125640010,125641000,125642500
compliance-icons compliance-icons
Product Identifier
  • Methanesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanesulfonyl chloride, 99.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.