missing translation for 'onlineSavingsMsg'
Learn More
Methanol with 0.1% Formic Acid, Honeywell™ Riedel-de-Haën™ CHROMASOLV™ Product Code.: 15621410

Methanol with 0.1% Formic Acid, Honeywell™ Riedel-de-Haën™ CHROMASOLV™

Product Code. 15621410
2 L, Glass Bottle
Click to view available options
Quantity:
2 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 15621410

Brand: Honeywell-Riedel-de Haen 142762L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CHROMASOLV™ LC-MS Ultra, LabReady™ Solvent Blend, tested for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles

 

TRUSTED_SUSTAINABILITY

Specifications

CAS 64-18-6, 67-56-1
Quantity 2 L
Packaging Glass Bottle
Molecular Formula CH2O2
MDL Number MFCD00004595
UN Number UN1230
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
SMILES OC=O
IUPAC Name formic acid
Molecular Weight (g/mol) 46.03
Formula Weight 32.04g/mol
Grade LC-MS Ultra CHROMASOLV™
Chemical Name or Material Methanol With 0.1% Formic Acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol with 0.1% formic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methanol, formic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanol with 0.1% Formic Acid, Honeywell™ Riedel-de-Haën™ CHROMASOLV™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.