missing translation for 'onlineSavingsMsg'
Learn More

Methanol with 0.1% Formic Acid (v/v), Certified HPLC Gradient Grade

Product Code. 12327083 Shop All Fisher Chemical Products
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Unit Size:
2.5 L
This item is not returnable. View return policy

Product Code. 12327083

Brand: Fisher Chemical FE/1300/17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 67-56-1
Quantity 2.5 L
Packaging Amber Glass Bottle
Molecular Formula CH4O
MDL Number MFCD00004595
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name formic acid
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol with 0,1% Formic acid (v/v) for HPLC Gradient analysis
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contains : Methyl alcohol, Formic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.