missing translation for 'onlineSavingsMsg'
Learn More

Methanol, 99.8%, Thermo Scientific Chemicals

For HPLC

£28.74 - £48.54

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10285131
View Documents Promotion Details
Thermo Scientific Acros
268280010
1 L Glass Bottle
£28.74
1 L
Please to purchase this item. Need a web account? Register with us today!
10124490
View Documents Promotion Details
Thermo Scientific Acros
268280025
2.5 L Glass Bottle
£48.54
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
Specifications

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO

Specifications

-98.0°C
67-56-1
0.0002 meq/g max.
0.7910g/mL
12°C
1.3280 to 1.3300
5ppm max.
CH4O
MFCD00004595
0.55 mPa.s (20°C)
0.05% Max.
15, 6029
Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
CO
32.04
CHEBI:17790
99.8%
Methanol, for HPLC
(1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 250nm, 0.005 max. at 260nm, 0.02 max. at 240nm, 0.05 max. at 230nm, 0.1 max. at 220nm, 0.2 max. at 210nm, 1 max. at 205nm
Colorless to Yellow
0.0002 meq/g max.
64.7°C
Liquid
99.8%
0.791
CH3OH
Vapor Pressure: 128hPa at 20°C
01, IV, 1227
11,322; 13,337
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
HPLC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

Danger

missing translation for 'einecsNumber' : 200-659-6

missing translation for 'rtecsNumber' : PC1400000

missing translation for 'tsca' : TSCA

Product Identifier
  • Methanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only