missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, ACS Spectrophotometric Grade, ≥99.9%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2 L
4 x 2 L
Packaging:
Glass Bottle
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
Melting Point | -98°C (literature) |
Absorbance | max. 0.01 at 400 nm, max. 0.01 280 nm, max. 0.04 at 260 nm, max. 0.10 at 240 nm, max. 0.20 at 230 nm, max. 0.40 at 220 nm, max. 0.80 at 210 nm, max. 1.00 at 205 nm |
CAS Max % | ≤100.0000% |
Density | 0.791 g/mL at 25 °C(lit.) |
Boiling Point | 64°C to 65°C |
Quantity | 2 L |
Explosion Limit | 0.36 |
Flash Point | 11°C |
Ignition Point | 725°F |
Packaging | Glass Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'