missing translation for 'onlineSavingsMsg'
Learn More

Methanol, ACS Spectrophotometric Grade, ≥99.9%, Honeywell Riedel-de Haën™

Product Code. p-7120572
Click to view available options
Quantity:
500 mL
1 L
2 L
4 x 2 L
Packaging:
Glass Bottle
Unit Size:
1 L
2 L
500mL
Pack of 4
This item is not returnable. View return policy

Product Code. 15691460

Brand: Honeywell-Riedel-de Haen 154903500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Spectrophotometric Grade,≥99.9%

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Density 0.791g/mL at 25°C
Boiling Point 64°C to 65°C
Quantity 500 mL
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Packaging Glass Bottle
Refractive Index 1.329
Linear Formula CH3OH
Vapor Pressure 97.68mmHg (20°C), 410mmHg (50°C)
UN Number UN1230
Beilstein 1098229
Formula Weight 32.04g/mol
Percent Purity ≥99.9%
Grade ACS
Chemical Name or Material Methanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.