missing translation for 'onlineSavingsMsg'
Learn More
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals Product Code.: 10327360

Methanol, 99.9%, for analysis, Thermo Scientific Chemicals

Product Code. 10327360
2.5 L
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
25 L
This item is not returnable. View return policy

Product Code. 10327360

Brand: Thermo Scientific Acros 176840025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spiritShow MoreShow Less
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98.0°C
Density 0.7910g/mL
Boiling Point 64.7°C
Quantity 2.5 L
Flash Point 12°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3280 to 1.3300
Assay Percent Range 99.9%
Residue after Evaporation 0.002% max.
Specific Gravity 0.791
Titratable Acid 0.0003 meq/g max.
Linear Formula CH3OH
Titratable Base 0.0002 meq/g max.
Viscosity 0.55 mPa.s (20°C)
Water 0.05% max. (K.F.)
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Formula Weight 32.04
Percent Purity 99.9%
Grade Analytical
Chemical Name or Material Methanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2524144 Certificate TypeCertificate of Analysis Date24/01/2025 Catalog Number176840100,176840025,176840250,176840010,176845000
Lot Number2417817 Certificate TypeCertificate of Analysis Date18/01/2025 Catalog Number176840100,176840025,176840250,176840010,176845000
Lot Number2522422 Certificate TypeCertificate of Analysis Date09/01/2025 Catalog Number176840100,176840025,176840250,176840010,176845000
Lot NumberBS24/089 Certificate TypeCertificate of Analysis Date24/12/2024 Catalog Number176840100,176840025,176840250,176840010,176845000
Lot Number2411951 Certificate TypeCertificate of Analysis Date23/12/2024 Catalog Number176840100,176840025,176840250,176840010,176845000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.