missing translation for 'onlineSavingsMsg'
Learn More
Methanol, 99%, Thermo Scientific Chemicals Product Code.: 11347996

Methanol, 99%, Thermo Scientific Chemicals

Product Code. 11347996
500 mL
Click to view available options
Quantity:
500 mL
1000 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11347996

Brand: Thermo Scientific Alfa Aesar L13255.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role in the measurement of methyl ester content of pectin. Further, it is widely used as a solvent in synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role in the measurement of methyl ester content of pectin. Further, it is widely used as a solvent in synthetic organic chemistry.

Solubility
Miscible with water and most of the organic solvents including ethyl acetate, toluene, dichloromethane, chloroform, tetrahydrofuran, and acetone.

Notes
Hygroscopic. Keep the material away from heat, sparks, flame and other sources of ignition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Density 0.791
Boiling Point 64.7°C
Quantity 500 mL
Flash Point 11°C (52°F)
Odor Mild
Refractive Index 1.329
UN Number UN1230
Beilstein 1098229
Merck Index 14,5957
Solubility Information Miscible with water and most of the organic solvents including ethyl acetate,toluene,dichloromethane,chloroform,tetrahydrofuran,and acetone.
Formula Weight 32.04
Percent Purity 99%
Chemical Name or Material Methanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2416864 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog NumberL13255.AU,L13255.AP,L13255.0F
Lot Number2400529 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL13255.AU,L13255.AP,L13255.0F
Lot NumberT29K012 Certificate TypeCertificate of Analysis Date06/08/2024 Catalog NumberL13255.AU,L13255.AP,L13255.0F
Lot NumberY14J006 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog NumberL13255.AU,L13255.AP,L13255.0F
Lot Number2487100 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog NumberL13255.AU,L13255.AP,L13255.0F
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Methanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.