missing translation for 'onlineSavingsMsg'
Learn More

Methanol, ACS Reagent, ≥99.8%, Honeywell Riedel-de Haën™

Product Code. 15653650
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.8%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Spezifikation

Melting Point -98°C (literature)
CAS 67-56-1
CAS Max % ≤100.0000%
Density 0.791 g/mL at 25 °C(lit.)
Boiling Point 64°C to 65°C
Quantity 1 L
Flash Point 11°C
Packaging Glass Bottle
Molecular Formula CH4O
Linear Formula CH3OH
MDL Number MFCD00004595
UN Number UN1230
Beilstein 1098229
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04
Percent Purity >99.8%
Grade ACS Reagent
Chemical Name or Material Methanol, ACS Reagent, ≥99.8%
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'