missing translation for 'onlineSavingsMsg'
Learn More
Methanol, ACS reagent, ≥99.8%, Honeywell Riedel-de Haën™ Product Code.: 15651610

Methanol, ACS reagent, ≥99.8%, Honeywell Riedel-de Haën™

Product Code. 15651610
2.5 L
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
6 x 500 mL
20 L
6 x 1 L
This item is not returnable. View return policy

Product Code. 15651610

Brand: Honeywell-Riedel-de Haen 1793372.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS reagent,≥99.8%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C (literature)
CAS Max % 100.0000%
Density 0.791 g/mL at 25 °C(lit.)
Boiling Point 64.7°C
Quantity 2.5 L
Flash Point 11°C
Packaging Glass Bottle
Linear Formula CH3OH
UN Number UN1230
Beilstein 1098229
Formula Weight 32.04
Percent Purity >99.8%
Grade ACS Reagent
Chemical Name or Material Methanol, ACS reagent, ≥99.8%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanol, ACS reagent, ≥99.8%, Honeywell Riedel-de Haën™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.