missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, ACS reagent, ≥99.8%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
6 x 500 mL
20 L
6 x 1 L
Unit Size:
1 L
2.5 L
20 L
4 L
500mL
Pack of 6
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Specifications
| Density | 0.791g/mL at 25°C |
| Quantity | 6 x 1 L |
| Explosion Limit | 0.36 |
| Flash Point | 9°C (48.2°F) |
| Ignition Point | 725°F |
| Linear Formula | CH3OH |
| Vapor Pressure | 410mmHg (50°C), 97.68mmHg (20°C) |
| UN Number | 1230 |
| Beilstein | 1098229 |
| Formula Weight | 32.04g/mol |
| Show More |
Safety and Handling
Product Identifier
- Methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction