missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, anhydrous, 99.9%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
Beschreibung
Methanol is widely used as a solvent in synthetic organic chemistry. It is also used as a solvent in analytical chemistry, especially for HPLC, LC-MS, GC-MS, ICP-MS and spectrophotometry. It is used in the production of biodiesel, formaldehyde and acetic acid. It is also used as fuel, antifreeze, denaturant for ethanol and an energy carrier for fuel cell application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Melting Point | °C to 98°C |
Density | 0.791 g/mL |
Boiling Point | 64.7°C |
Quantity | 250 mL |
Flash Point | 11°C (52°F) |
Physical Form | Liquid |
Refractive Index | 1.329 |
Assay Percent Range | 99.9% |
Beilstein | 1098229 |
Solubility Information | Miscible with water and most of the organic solvents including ethyl acetate,toluene,dichloromethane,chloroform,tetrahydrofuran,and acetone. |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'