missing translation for 'onlineSavingsMsg'
Learn More

Methanol, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10281001
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Exceeds ACS Specifications

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Hydroxy methane , Methyl alcohol , Carbinol , Wood alcohol
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Max. 10 (APHA)
Appearance Clear
Density 0.7918 g/cm³ (20°C)
Boiling Point 64.6°C (1013 hPa)
Quantity 1 L
Flash Point 11°C
Packaging Glass Bottle
Assay Percent Range min. 99.8 % (by GC, corrected for water)
Residue after Evaporation max. 0.001 %
Water max. 0.05 %
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'