missing translation for 'onlineSavingsMsg'
Learn More
Methanol, BAKER ANALYZED™ ULTRA LC/MS, J.T. Baker™ Product Code.: 15588664
compliance-icons compliance-icons compliance-icons

Methanol, BAKER ANALYZED™ ULTRA LC/MS, J.T. Baker™

Product Code. 15588664
6 x 1 L, Narrow Mouth Flint
Click to view available options
Quantity:
6 x 1 L
Packaging:
Narrow Mouth Flint
This item is not returnable. View return policy

Product Code. 15588664

Brand: Avantor J.T.Baker 986302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cutting-edge LC/MS applications

    For Use in LC/MS and UHPLC; For Laboratory, Research, or Manufacturing Use

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point 113°C to 116°C
Quantity 6 x 1 L
Flash Point 12.00°C
Packaging Narrow Mouth Flint
Specific Gravity 1L = 0.79kg
Formula Weight 32.04
Grade BAKER ANALYZED™ ULTRA LC/MS
Chemical Name or Material Methanol
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanol, BAKER ANALYZED™ ULTRA LC/MS, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.