missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, For pesticide residue analysis, Honeywell Riedel-de Haën™
CHROMASOLV™, for pesticide residue analysis
£38.31 - £277.55
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15634200
|
Honeywell-Chromasolv
34485-1L |
1 L |
£38.31
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15644200
|
Honeywell-Chromasolv
34485-2.5L |
2.5 L |
£79.51
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15654200
|
Honeywell-Chromasolv
34485-4X2.5L |
4 x 2.5 L |
£277.55
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
67-56-1 | |
0.36 | |
725°F | |
CH3OH | |
410 mmHg (50°C), 97.68 mmHg (20°C) | |
1098229 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04g/mol | |
CHROMASOLV™ |
0.791g/mL at 25°C | |
9°C (48.2°F) | |
CH4O | |
MFCD00004595 | |
UN1230 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
≥99.9% (GC) | |
Methanol |
Safety and Handling
P210-P260-P280-P301 + P310-P311
missing translation for 'rtecsNumber' : PC1400000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Methanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methanol, For pesticide residue analysis, Honeywell Riedel-de Haën™