missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Dried, BAKER ANALYZED, J.T. Baker™

Product Code. 10149610 Shop All J.T.Baker Products
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Unit Size:
2.5 L
This item is not returnable. View return policy

Product Code. 10149610

Brand: JT Baker 9049.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality grade chemical that exceeds ACS specifications for laboratory and specialized industrial use, with a maximum of 0.01% H2O.

  • Exceeds the requirements set forth by the American Chemical Society (ACS)
  • Meets Reagent Specificatons for USP/NF for use in pharmaceutical processing and prescription compounding
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Melting Point -97.8°C
CAS 67-56-1
Color Colorless
Appearance Clear
Density 0.79 g/mL at 25°C
Boiling Point 64°C
Quantity 2.5 L
Explosion Limit 6 - 36 Vol %
Flash Point 11°C
Ignition Point 240°C
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 0.001%
Molecular Formula CH4O
Titratable Acid Max. 0.0003meq/g
Linear Formula CH3OH
Titratable Base Max. 0.0002meq/g
MDL Number MFCD00004595
Vapor Pressure 16.9 kPa
Water Max. 0.01%
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Solubility Information Miscible in ether, benzene, ethanol; Soluble in acetone, chloroform
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04g/mol
Percent Purity ≥99.8%
Grade Exceeds ACS specifications
Chemical Name or Material Methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.