missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Extra Dry, for Synthesis, Fisher Chemical™
CH4O, CAS Number-67-56-1, methyl hydroxide, methylol, pyroxylic spirit, carbinol, wood spirit, wood naphtha, wood alcohol, colonial spirit, columbian spirit, methyl alcohol, 1L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042
£49.95 - £555.00
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10020260
|
Fisher Chemical
M/4050/08 |
500 mL | Amber Glass Bottle |
£49.95
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10468410
|
Fisher Chemical
M/4050/15 |
1 L | Amber Glass Bottle |
£74.60
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10598240
|
Fisher Chemical
M/4050/17 |
2.5 L | Amber Glass Bottle |
£118.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10366140
|
Fisher Chemical
M/4050/PB17 |
2.5 L | Plastic-coated Glass Bottle |
£153.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10745721
|
Fisher Chemical
M/4050/25 |
25 L | Metal Can |
£555.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CH4O, CAS Number-67-56-1, methyl hydroxide, methylol, pyroxylic spirit, carbinol, wood spirit, wood naphtha, wood alcohol, colonial spirit, columbian spirit, methyl alcohol, 1L, 64.7 deg.C, CHEBI:17790, Colorless, 32.04g/mol, OKKJLVBELUTLKV-UHFFFAOYSA-N, methanol, -98 deg.C, 4595, 32.042
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
-98°C | |
Colorless | |
Liquid | |
MFCD00004595 | |
0.55cp at 20°C | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04g/mol |
67-56-1 | |
64.7°C | |
CH4O | |
128 hPa at 20°C | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Methanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P240-Ground/bond container and receiving equipment.