missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Laboratory Reagent, ≥99.6%, Honeywell™

Product Code. p-7120577 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
25 L
4 x 2.5 L
4 x 4 L
4 x 5 L
160 kg
Packaging:
Fiberboard Box
Glass Bottle
Metal Drum
Stainless Steel Can
Förpackningsstorlek
1 L
10 L
160kg
2.5 L
20 L
25 L
Pack of 4
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 15601630

missing translation for 'mfr': Honeywell Research Chemicals 1799571L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Laboratory Reagent,≥99.6%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifikationer

Melting Point -98°C
Color Colorless
Density 0.791g/mL at 25°C
Boiling Point 65°C
Quantity 1 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.329(lit.)
Linear Formula CH3OH
Vapor Pressure 97.68mmHg (20°C), 410mmHg (50°C)
UN Number UN1230
Beilstein 1098229
Formula Weight 32.04g/mol
Percent Purity ≥99.6%
Grade Laboratory Reagent
Chemical Name or Material Methanol
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • Methanol
Signalord
  • Danger
Riskkategorier
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Faroangivelser
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H370-Causes damage to organs.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.