missing translation for 'onlineSavingsMsg'
Learn More

Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™

£42.90 - £68.00

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10163383
View Documents
Fisher BioReagents
BP1105-1
1 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10785484
View Documents Promotion Details
Fisher BioReagents
BP1105-4
4 L Glass Bottle
£68.00
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Methanol (Peroxide-Free/Sequencing)

Methanol is used in staining and destaining protein electrophoresis gels, in HPLC, and in other biological applications.
Specifications

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO

Specifications

-98°C
67-56-1
0.791g/cm³
Liquid
≥99.9 %
CH4O
CH3OH
MFCD00004595
0.55 cP at 20°C
15, 6029
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
Peroxides: ≤0001%
0.025 max. at 254nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 1 max. at 205nm
Colorless
64.7°C
1.326 to 1.330 (at 25°C)
3ppm max.
0.0003mEq/g max.
0.0002mEq/g max.
128 hPa at 20°C
0.1% max.
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.04
CHEBI:17790
≥99.9%
Methanol, Peroxide-free
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0

missing translation for 'dotInformation' : DOT Class 3, : Flammable Liquid

Recommended Storage : RT

Product Identifier
  • Methanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.

SDS
Documents

Documents

Product Certifications