missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
45 kg
150 kg
Specifications
Specifications
Density | 0.791g/mL at 25°C |
Quantity | 45 kg |
Explosion Limit | 0.36 |
Flash Point | 9°C (48.2°F) |
Ignition Point | 725°F |
Linear Formula | CH3OH |
Vapor Pressure | 410mmHg (50°C), 97.68mmHg (20°C) |
UN Number | 1230 |
Beilstein | 1098229 |
Formula Weight | 32.04g/mol |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 14
Safety and Handling



Product Identifier
- Methanol
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8% (GC), Honeywell Riedel-de Haën™ >
Spot an opportunity for improvement?Share a Content Correction