missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8% (GC), Honeywell Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.8% (GC)
£158.64 - £402.70
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15693710
|
Honeywell-Riedel-de Haen
32213-6X1L |
6 x 1 L | Poly Bottle |
£158.64
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15663710
|
Honeywell-Riedel-de Haen
32213-4X2.5L |
4 x 2.5 L | Poly Bottle |
£233.60
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15673710
|
Honeywell-Riedel-de Haen
32213-4X5L |
4 x 5 L | Poly Bottle |
£402.70
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
-98°C | |
Colorless | |
0.791g/mL at 25°C | |
0.36 | |
725°F | |
0 to 0.0005 % | |
Free acid (as HCOOH): ≤0.002% | |
Free Alkali (as NH3): ≤0.00005% | |
410mmHg (50°C), 97.68mmHg (20°C) | |
1098229 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04g/mol | |
ACS/puriss. p.a. |
67-56-1 | |
Liquid | |
64°C to 65°C | |
9°C (48.2°F) | |
1.328 to 1.33 | |
CH4O | |
CH3OH | |
MFCD00004595 | |
UN1230 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
≥99.8% (GC) | |
Methanol, Reag. Ph. Eur. reag. ISO |
Safety and Handling
P210-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378-P403 + P235
H225-H301 + H311 + H331-H370
missing translation for 'dotInformation' : Class 3 - PG 2 - Methanol
missing translation for 'einecsNumber' : 200-659-6
missing translation for 'rtecsNumber' : PC1400000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Methanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Methanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.8% (GC), Honeywell Riedel-de Haën™