missing translation for 'onlineSavingsMsg'
Learn More

Methanol, Technical, Fisher Chemical™

£55.40 - £358.00

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10606652
SDS View Documents Promotion Details
Fisher Chemical
M/3900/17
2.5 L Amber Glass Bottle
£55.40
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10508060
SDS View Documents Promotion Details
Fisher Chemical
M/3900/PB17
2.5 L Plastic-coated Glass Bottle
£70.90
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10121720
SDS View Documents Promotion Details
Fisher Chemical
M/3900/25
25 L Metal Can
£358.00
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanol, Technical, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.