missing translation for 'onlineSavingsMsg'
Learn More
Methanol, technical Product Code.: 13218003

Methanol, technical

Product Code. 13218003
5 L, Plastic Drum
Click to view available options
Quantity:
5 L
Packaging:
Plastic Drum
This item is not returnable. View return policy

Product Code. 13218003

Brand: Thermo Scientific Acros 444310050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67-56-1 | CH4O | 32.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -98.0°C
CAS 67-56-1
Density 0.7910g/mL
Boiling Point 64.7°C
Quantity 5 L
Flash Point 12°C
Packaging Plastic Drum
Refractive Index 1.3280 to 1.3300
Specific Gravity 0.791
Molecular Formula CH4O
Linear Formula CH3OH
MDL Number MFCD00004595
Viscosity 0.55 mPa.s (20°C)
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04
Grade Technical
Chemical Name or Material Methanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466024 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog Number444310050
Lot NumberA0459174 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog Number444310050
Lot NumberA0455204 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number444310050
Lot NumberA0392812 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number444310050
Lot NumberA0398616 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number444310050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methanol, technical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.