Learn More
Methanol, UHPLC-MS, Thermo Scientific™
Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer.
Brand: Thermo Scientific A458-1
100.25 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 67-56-1 Weight : 0.79100kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Description
Methanol, CH4O, CAS Number-67-56-1, wood naphtha, pyroxylic spirit, methyl alcohol, methylol, columbian spirit, colonial spirit, wood spirit, wood alcohol, methyl hydroxide, carbinol, 1L
Specifications
Methanol | |
67-56-1 | |
Colorless | |
64.7°C | |
Clear Borosilicate Glass Bottle | |
1ppm max. | |
0.0003meq/g max. | |
0.0002meq/g max. | |
MFCD00004595 | |
0.55 cP at 20°C | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 |
-98°C | |
≤0.005 AU at 280nm, ≤0.01 AU at 254nm, ≤0.1 AU at 230nm, ≤0.4 AU at 215nm, ≤000 5 AU at 260nm, ≤02. AU at 220nm, ≤05. AU at 210nm | |
0.791 g/cm3 | |
1 L | |
Liquid | |
99.9% minimum by GC | |
CH4O | |
CH3OH | |
128 hPa at 20°C | |
0.02% Max. | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
UHPLC-MS |
Safety and Handling
- Methanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only