missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Methionine Sulfoxide Reductase A Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Brand: Novus Biologicals™ NBP2-55671PEP
Additional Details : Weight : 0.00970kg
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human Methionine Sulfoxide Reductase A. Source: E.coli Amino Acid Sequence: AKQMEAALSSKENYQKVLSEHGFGPITTDIREGQTFYYAEDYHQQYLSKNPNGYCGLGGTGVSCPVGIK The Methionine Sulfoxide Reductase A Recombinant Protein Antigen is derived from E. coli. The Methionine Sulfoxide Reductase A Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.Specifications
4482 | |
Methionine Sulfoxide Reductase A Recombinant Protein Antigen | |
PBS and 1M Urea, pH 7.4. | |
cytosolic methionine-S-sulfoxide reductase, EC 1.8.4.11, methionine sulfoxide reductase A, peptide met (O) reductase, Peptide Met(O) reductase, peptide methionine sulfoxide reductase, Peptide-methionine (S)-S-oxide reductase, PMSR, Protein-methionine-S-ox | |
Unlabeled | |
100μL | |
E.Coli |
>80% by SDS-PAGE and Coomassie blue staining | |
Store at −20C. Avoid freeze-thaw cycles. | |
Blocking/Neutralizing, Control | |
MSRA | |
Recombinant Protein Antigen | |
RUO | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-51825. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
For Research Use Only