Learn More
Methoprene, Honeywell Fluka™
Brand: Honeywell Fluka R1783-100MG
Additional Details : Weight : 0.10000kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Methoprene | |
100 mg | |
Beige | |
1913191 | |
310.5 g/mol | |
100°C (0.05 hPa) |
40596-69-8 | |
≤ 100% | |
C19H34O3 | |
isopropyl (E,E)-(RS)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate | |
0.921 g/cm3 (25°C) | |
136°C |