Learn More
Thermo Scientific™ Methoxamine (MOX) Reagent
Prepare oximes of steroids and ketoacids prior to silylation with the Thermo Scientific™ Methoxamine (MOX) Reagent.
Brand: Thermo Scientific™ TS-45950
80.91 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Prepare oximes of steroids and ketoacids prior to silylation with the Thermo Scientific™ Methoxamine (MOX) Reagent. It prevents the formation of multiple derivatives when enols are present during silylation, thereby ensuring optimal quantitation. The reagent is a 2% solution of methoxyamine–hydrogen chloride in pyridine.
- 2% methoxyamine·HCl (M.W. 83.51) in pyridine
- Prevents formation of multiple derivatives when enols are present during silylation
- Supplied in amber Thermo Scientific Hypo-Vial Sample Storage Vial with septum and crimp top
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Preparing Oximes of Steroids and Ketoacids | |
95 - 98, 1 - 3% | |
2°C to 8°C |
Corrosive, Flammable | |
Hypo-Vial Container; 10 mL | |
Pyridine, Methoxyammonium chloride |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- MOX Reagent, 10 mL
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P370+P378-In case of fire: Use to extinguish.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Pyridine
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.