missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Methoxycarbonylmethylene)triphenylphosphorane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
(Methoxycarbonylmethylene)triphenylphosphorane is used in olefination reactions. Further, it undergoes the Wittig reaction with aldehydes to give substituted methyl acrylates. It is used in the preparation of (triphenylphosphoranylidene)-ketene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(Methoxycarbonylmethylene)triphenylphosphorane is used in olefination reactions. Further, it undergoes the Wittig reaction with aldehydes to give substituted methyl acrylates. It is used in the preparation of (triphenylphosphoranylidene)-ketene.
Solubility
Soluble in chloroform. Slightly soluble in ethanol and terahydrofuran. Insoluble in water.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
(Methoxycarbonylmethylene)triphenylphosphorane is used in olefination reactions. Further, it undergoes the Wittig reaction with aldehydes to give substituted methyl acrylates. It is used in the preparation of (triphenylphosphoranylidene)-ketene.
Solubility
Soluble in chloroform. Slightly soluble in ethanol and terahydrofuran. Insoluble in water.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | 165°C to 169°C |
Quantity | 25 g |
Beilstein | 618430 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in chloroform. Slightly soluble in ethanol and terahydrofuran. Insoluble in water. |
Formula Weight | 334.36 |
Percent Purity | 98% |
Chemical Name or Material | (Methoxycarbonylmethylene)triphenylphosphorane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'