missing translation for 'onlineSavingsMsg'
Learn More

Methoxycarbonylsulfenyl chloride, 95%, Thermo Scientific Chemicals

Product Code. 11386896
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Methoxycarbonylsulfenyl chloride is used to prepare 3-[(methoxycarbonyl)dithio]-L-alanine by reacting with L-cysteine. Further, it is used in the synthesis of 3-2 pyridinyldithio propanoic acid hydrazide (PDPH). It serves as an important heterobifunctional crosslinker, which is a crosslinker used to make macromolecule-drug conjugates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxycarbonylsulfenyl chloride is used to prepare 3-[(methoxycarbonyl)dithio]-L-alanine by reacting with L-cysteine. Further, it is used in the synthesis of 3-2 pyridinyldithio propanoic acid hydrazide (PDPH). It serves as an important heterobifunctional crosslinker, which is a crosslinker used to make macromolecule-drug conjugates.

Solubility
Miscible with dichlormethane.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26555-40-8
Molecular Formula C2H3ClO2S
Molecular Weight (g/mol) 126.554
MDL Number MFCD00013648
InChI Key TXJXPZVVSLAQOQ-UHFFFAOYSA-N
Synonym methoxycarbonylsulfenyl chloride, methoxycarbonyl sulfenyl chloride, s-chloro o-methyl thiocarbonate, thiocarbonic acid, anhydrosulphide with methyl thiohypochlorite, carbonothioic acid, anhydrosulfide with thiohypochlorous acid, methyl ester, acmc-209gs8, methoxycarbonylsulfenylchloride, chlorothio methoxy oxomethane, methoxy carbonylsulfenyl chloride, methoxy-carbonylsulfenyl chloride
PubChem CID 123417
IUPAC Name methyl chlorosulfanylformate
SMILES COC(=O)SCl

Specifications

Density 1.398
Boiling Point 73°C to 74°C (100 mmHg)
Flash Point 54°C (129°F)
Refractive Index 1.482
Quantity 5 g
UN Number UN2920
Beilstein 1849908
Sensitivity Moisture sensitive
Solubility Information Miscible with dichlormethane.
Formula Weight 126.56
Percent Purity 95%
Chemical Name or Material Methoxycarbonylsulfenyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.