missing translation for 'onlineSavingsMsg'
Learn More

Methoxychlor, Thermo Scientific Chemicals

Product Code. 15465999
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Molecule with estrogenic activity

Molecule with estrogenic activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-43-5
Molecular Formula C16H15Cl3O2
Molecular Weight (g/mol) 345.644
MDL Number MFCD00000803
InChI Key IAKOZHOLGAGEJT-UHFFFAOYSA-N
Synonym methoxychlor, p,p'-methoxychlor, dmdt, dimethoxy-ddt, methoxy-ddt, methoxcide, metox, maralate, marlate, moxie
PubChem CID 4115
ChEBI CHEBI:6842
IUPAC Name 1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene
SMILES COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)C(Cl)(Cl)Cl

Specifications

Melting Point 87°C
Color White
Quantity 25 g
UN Number UN3077
Beilstein 2057367
Merck Index 14,5990
Formula Weight 345.65
Physical Form Crystalline Powder
Chemical Name or Material Methoxychlor

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'